Would you like to log in again?
Lenses and Detectors for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: Agilent
8003-0564 8003-0566 8003-0566 8003-0564
AG8003-0566EA 141.72 USD InStock
AG8003-0566 CAAG8003-0564 CAAG8003-0566 AG8003-0564
Lenses and Detectors for PerkinElmer ICP-MS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Ion lenses for PerkinElmer ELAN & SimulScan detectors for PerkinElmer NexION 300/350 & ELAN ICP-MS.
  • Guaranteed to be compatible with PerkinElmer ICP-MS instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent ion lenses and SimulScan detectors are equivalent to the PerkinElmer ICP-MS versions and can be used with complete confidence.;Guaranteed to be compatible with PerkinElmer ICP-MS instruments and backed by the Agilent Stand Behind Warranty that additionally guarantees the parts will not cause instrument failure or downtime.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty