Would you like to log in again?
CrossLab Adapters and Connectors for PerkinElmer FIMS/FIAS, Agilent Technologies
Supplier: Agilent
8003-0592 8003-0667 8003-0675 8003-0657 8003-0592 8003-0654 8003-0657 8003-0659 8003-0667 8003-0664 8003-0664 8003-0662 8003-0658 8003-0662 8003-0658 8003-0659 8003-0663 8003-0654 8003-0675
AG8003-0592EA 6.46 USD InStock
CAAG8003-0592 AG8003-0592 AG8003-0663 AG8003-0662 AG8003-0667 CAAG8003-0659 AG8003-0654 AG8003-0664 AG8003-0675 AG8003-0659 AG8003-0658 AG8003-0657 CAAG8003-0662 CAAG8003-0654 CAAG8003-0675 CAAG8003-0664 CAAG8003-0667 CAAG8003-0658 CAAG8003-0657
CrossLab Adapters and Connectors for PerkinElmer FIMS/FIAS, Agilent Technologies
Fittings and Connectors
Agilent adapters with internal or external Thread and connectors for PerkinElmer FIMS/FIAS systems.
  • Guaranteed to be compatible with PerkinElmer FIMS/FIAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent adapters with internal thread, adapters with external thread, and connectors for PerkinElmer FIMS/FIAS systems are equivalent to genuine adapters and connectors for PerkinElmer FIMS/FIAS systems. You can be confident that these adapters will integrate seamlessly with your PerkinElmer instrument.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty