Would you like to log in again?
CrossLab Tubing and Tubing Assemblies for PerkinElmer FIMS/FIAS, Agilent Technologies
Supplier: Agilent
8003-0688 8003-0682 8003-0697 8003-0686 8003-0687 8003-0682 8003-0411 8003-0412 8003-0685 8003-0412 8003-0696 8003-0686 8003-0690 8003-0687 8003-0695 8003-0700 8003-0683 8003-0685 8003-0684 8003-0700 8003-0690 8003-0695 8003-0691 8003-0684 8003-0688 8003-0696 8003-0683 8003-0697 8003-0691
AG8003-0685PK 110.83 USD InStock
CAAG8003-0683 CAAG8003-0682 AG8003-0685 AG8003-0684 AG8003-0683 AG8003-0682 AG8003-0700 AG8003-0688 AG8003-0687 AG8003-0686 CAAG8003-0685 CAAG8003-0684 CAAG8003-0687 CAAG8003-0686 CAAG8003-0700 CAAG8003-0688 CAAG8003-0690 CAAG8003-0691 AG8003-0691 AG8003-0690 AG8003-0696 AG8003-0695 AG8003-0412 AG8003-0411 AG8003-0697 CAAG8003-0696 CAAG8003-0695 CAAG8003-0412 CAAG8003-0697
CrossLab Tubing and Tubing Assemblies for PerkinElmer FIMS/FIAS, Agilent Technologies
Tubing Chromatography Tubing
Agilent tubing and tubing assemblies are fully compatible with your PerkinElmer FIMS/FIAS systems.
  • Guaranteed to be compatible with PerkinElmer FIMS/FIAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
Agilent tubing and tubing assemblies are fully compatible with your PerkinElmer FIMS/FIAS systems, so you can be confident that they will integrate seamlessly with your PerkinElmer system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty