Would you like to log in again?
O-Rings and Supplies for PerkinElmer AAS Nebulizers and Burner Systems
Supplier: Agilent
CrossLab
8003-0835 8003-0853 8003-0862 8003-0863 8003-0843 8003-0853 8003-0861 8003-0849 8003-0835 8003-0834 8003-0863 8003-0844 8003-0397 8003-0842 8003-0852 8003-0862 8003-0826 8003-0861 8003-0825 8003-0826 8003-0849 8003-0834 8003-0843 8003-0397 8003-0852 8003-0825
AG8003-0861EA 12.67 USD InStock
AG8003-0861 AG8003-0843 CAAG8003-0825 AG8003-0842 AG8003-0863 CAAG8003-0849 AG8003-0862 CAAG8003-0826 AG8003-0825 AG8003-0844 CAAG8003-0861 CAAG8003-0863 AG8003-0849 CAAG8003-0862 AG8003-0826 CAAG8003-0843 CAAG8003-0397 AG8003-0397 CAAG8003-0835 AG8003-0853 AG8003-0852 AG8003-0835 AG8003-0834 CAAG8003-0852 CAAG8003-0853 CAAG8003-0834
O-Rings and Supplies for PerkinElmer AAS Nebulizers and Burner Systems
Chromatography Supplies Chromatography Spare Parts
Agilent O-rings and supplies for PerkinElmer PinAAcle/Aanalyst flame atomic absorption systems.
  • Qualified on a genuine PerkinElmer system
  • Guaranteed to be compatible with PerkinElmer AA instruments
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
Agilent O-rings and supplies for PerkinElmer PinAAcle/Aanalyst flame atomic absorption systems are equivalent to genuine supplies for PerkinElmer PinAAcle/Aanalyst AA systems. You can be confident that these supplies will integrate seamlessly with your PerkinElmer instrument.
Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime. Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies. Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty