Would you like to log in again?
Nebulizers and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Supplier: Agilent
8003-0833 8003-0409 8003-0833 8003-0406 8003-0836 8003-0864 8003-0827 8003-0837 8003-0860 8003-0837 8003-0860 8003-0823 8003-0864 8003-0830 8003-0406 8003-0823 8003-0830 8003-0409 8003-0831 8003-0827 8003-0836 8003-0831
AG8003-0860EA 258.92 USD InStock
AG8003-0860 CAAG8003-0836 AG8003-0864 CAAG8003-0406 AG8003-0831 CAAG8003-0409 CAAG8003-0827 AG8003-0830 CAAG8003-0837 AG8003-0836 AG8003-0406 AG8003-0823 AG8003-0833 CAAG8003-0860 CAAG8003-0830 AG8003-0409 AG8003-0827 AG8003-0837 CAAG8003-0864 CAAG8003-0831 CAAG8003-0823 CAAG8003-0833
Nebulizers and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent nebulizers and supplies are equivalent to PerkinElmer AA nebulizer components.
  • Guaranteed to be compatible with PerkinElmer AA instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Nebulizers are a critical part of the Flame AA sample introduction system. Agilent nebulizers for PerkinElmer AA instruments are designed to support a variety of samples and provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty