Would you like to log in again?
Tools and Supplies for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: Agilent Technologies
8003-0955 8003-0423 8003-0423 8003-0414 8003-0955 8003-0956 8003-0414 8003-0956
AG8003-0414EA 134 USD InStock
AG8003-0414 CAAG8003-0956 AG8003-0423 AG8003-0956 AG8003-0955 CAAG8003-0423 CAAG8003-0414 CAAG8003-0955
Tools and Supplies for PerkinElmer GFAAS Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Tools for positioning the L'vov platform in the HGA graphite tube, and other supplies PE systems.
  • Guaranteed to be compatible with PerkinElmer Graphite Furnace AAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
These tools and supplies for PerkinElmer graphite furnace AAS systems can assist you with positioning the L'vov platform precisely in the center of the HGA graphite tube. These tools and supplies, include other supplies used with the PerkinElmer Graphite Furnace AA systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty