Would you like to log in again?
Mixing/Separation Assemblies for PerkinElmer FIMS/FIAS, Agilent Technologies
Supplier: Agilent Technologies
8003-0972 8003-0972 8003-0674 8003-0673 8003-0673 8003-0674
AG8003-0972EA 232 USD InStock
AG8003-0972 CAAG8003-0674 CAAG8003-0673 CAAG8003-0972 AG8003-0674 AG8003-0673
Mixing/Separation Assemblies for PerkinElmer FIMS/FIAS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent Mixing/Separation Assembly, Mix Block and Gas/Liq Separator for PerkinElmer FIMS/FIAS system.
  • Guaranteed to be compatible with PerkinElmer FIMS/FIAS systems;Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system;Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent mixing/separation assembly, mixing block and gas/liquid separator for PerkinElmer FIMS/FIAS systems are equivalent to genuine mixing/separation assembly, mixing block and gas/liquid separator for PerkinElmer FIMS/FIAS systems. With these supplies you can be confident that it will integrate seamlessly with your PerkinElmer instrument.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty