Would you like to log in again?
CrossLab Column Ferrules for Third Party GC Instruments
Supplier: Agilent
CrossLab
8003-0223 8001-0212 8001-0213 8003-0223 8004-0217 8001-0223 8002-0212 8010-0302 8004-0212 8010-0301 8010-0303 8003-0216 8010-0267 8002-0213 8010-0312 8001-0221 8001-0211 8010-0311 8002-0216 8004-0211 8002-0217 8010-0310 8003-0212 8002-0214 8004-0213 8010-0302 8010-0304 8001-0223 8002-0213 8010-0315 8004-0215 8004-0216 8010-0267 8010-0315 8001-0222 8010-0308 8002-0215 8010-0307 8004-0217 8004-0211 8010-0313 8002-0220 8010-0310 8002-0212 8001-0212 8002-0215 8010-0309 8004-0219 8004-0214 8010-0308 8010-0313 8010-0304 8004-0216 8004-0213 8003-0219 8010-0314 8010-0305 8010-0269 8002-0221 8010-0312 8010-0269 8010-0309 8002-0222 8003-0212 8001-0213 8010-0314 8010-0306 8010-0305 8010-0301 8004-0214 8003-0216 8002-0217 8001-0211 8004-0219 8010-0268 8010-0268 8003-0219 8001-0222 8002-0214 8010-0307 8010-0306 8010-0303 8004-0215 8010-0311 8001-0221 8002-0216 8002-0220 8002-0222 8004-0212
AG8003-0223PK 67.57 USD InStock
CAAG8002-0214 CAAG8002-0215 CAAG8002-0216 CAAG8002-0217 CAAG8002-0212 CAAG8002-0213 AG8003-0223 CA8004-0214 CA8004-0216 CAAG8003-0223 AG8002-0216 AG8002-0217 AG8002-0214 AG8002-0215 AG8002-0212 CA8004-0212 AG8002-0213 CA8010-0304 CAAG8010-0267 CAAG8002-0222 CAAG8010-0301 CA8010-0269 CAAG8010-0268 CAAG8010-0302 CAAG8004-0217 CAAG8010-0303 CAAG8010-0305 CAAG8004-0219 CAAG8001-0213 CAAG8010-0306 CAAG8010-0307 CAAG8001-0212 CAAG8004-0213 AG8003-0212 CAAG8001-0211 CAAG8010-0308 CAAG8004-0215 CAAG8010-0309 AG8003-0216 CAAG8004-0211 AG8003-0219 CAAG8002-0220 CAAG8010-0310 CAAG8010-0311 CAAG8010-0312 CAAG8010-0313 AG8001-0223 CAAG8010-0314 CAAG8010-0315 AG8001-0222 CAAG8001-0223 CAAG8001-0222 CAAG8001-0221 AG8001-0221 AG8010-0311 AG8010-0310 AG8010-0315 AG8010-0314 AG8010-0313 AG8010-0312 AG8002-0221 AG8002-0222 AG8002-0220 AG8001-0212 AG8004-0214 AG8001-0211 AG8004-0215 AG8004-0216 AG8004-0217 AG8001-0213 AG8004-0219 AG8010-0308 CAAG8003-0219 AG8010-0307 AG8010-0306 AG8010-0305 AG8004-0211 AG8004-0212 AG8004-0213 AG8010-0309 CAAG8003-0212 AG8010-0267 CAAG8003-0216 AG8010-0304 AG8010-0303 AG8010-0302 AG8010-0269 AG8010-0301 AG8010-0268
CrossLab Column Ferrules for Third Party GC Instruments
Fittings and Connectors
Column ferrules for PerkinElmer, Thermo Scientific, and Varian/Bruker GC Systems.
  • All column ferrules are nonstick and provide a leak-free seal
  • Graphite ferrules provide an easy-to-use and highly stable seal, boast a high temperature limit, and can be removed easily
  • Vespel/graphite ferrules are mechanically robust and maintain effectiveness over a long lifetime
  • Vespel ferrules are mechanically robust, maintain a long lifetime, and can be reused or removed easily
  • To ensure performance and longevity, column ferrules should be replaced at the same time as columns, and when performing column maintenance
Agilent provides a range of high-performance column ferrules for various third-party gas chromatography instruments, including PerkinElmer, Thermo Scientific and Varian/Bruker. Graphite, Vespel/graphite, and Vespel column ferrules create consistent, leak-free seals with minimum torque. All column ferrules accommodate various column outer diameters and can withstand temperature cycling. In addition, the specially designed column ferrules prevent sticking to columns or other fittings. Whatever your application, Agilent's column ferrule portfolio can deliver excellent performance for use with your GC system
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty