Would you like to log in again?
CrossLab Pump Supplies for Waters LC Systems, Agilent Technologies
Supplier: Agilent
CrossLab
8005-0513 8005-0513
AG8005-0513PK 403.86 USD InStock
CAAG8005-0513 AG8005-0513
CrossLab Pump Supplies for Waters LC Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies for maintaining selected Waters HPLC systems, including common pump replacement parts.
  • Agilent CrossLab parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent
Agilent pump supplies for selected Waters HPLC systems contain the most important components of a pump: plungers, seals, and check valves. Agilent HPLC supplies are manufactured and rigorously tested to ensure performance with your analytical instrument.
Regular pump maintenance helps lower operating costs and ensure precise and consistent results. By following a regular maintenance routine, you can count on maximum uptime, steady and accurate solvent flow, and pressure stability throughout the lifetime of the pump.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty