Would you like to log in again?
CrossLab Capillaries for Waters LC Systems, Agilent Technologies
Supplier: Agilent
8005-0812 5500-1261 5500-1260 8005-0822 5067-6210 8005-0824 5500-1262 8005-0823 8005-0826 5500-1263 5500-1259 5500-1261 8005-0812 5500-1262 8005-0826 8005-0824 8005-0822 5500-1260 5500-1259 5500-1263 8005-0823 5067-6210
AG5500-1259EA 79.18 USD InStock
CAAG5500-1263 CAAG8005-0824 CAAG5500-1262 CAAG5500-1261 CAAG8005-0822 CAAG8005-0812 CAAG5500-1260 CAAG8005-0823 CAAG8005-0826 CA5067-6210 CAAG5500-1259 AG5500-1259 AG5067-6210 AG5500-1263 AG8005-0824 AG5500-1262 AG8005-0826 AG5500-1261 AG8005-0812 AG8005-0823 AG8005-0822 AG5500-1260
CrossLab Capillaries for Waters LC Systems, Agilent Technologies
Tubing Chromatography Tubing
Stainless steel capillaries and tubing for selected Waters HPLC systems.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
  • Available in pre-swaged
Agilent offers a range of capillaries and tubing made from stainless steel for selected Waters HPLC systems. Used with the right fittings, they are ideal for LC system plumbing, tight, leak-free connections, and zero dead volumes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty