Would you like to log in again?
Liner O-Rings for Third Party GC Instruments, Agilent Technologies
Supplier: Agilent
8003-0202 8003-0205 8002-0201 8001-0202 8003-0205 8010-0401 8001-0203 8004-0201 8004-0204 8001-0201 8002-0203 8004-0202 8002-0201 8004-0202 8003-0202 8001-0203 8001-0201 8002-0204 8003-0203 8004-0201 8004-0203 8002-0204 8003-0204 8002-0203 8003-0204 8010-0401 8004-0203 8001-0202 8004-0204 8003-0203
AG8001-0201PK 30.85 USD InStock
AG8001-0201 AG8004-0203 AG8004-0204 CAAG8001-0203 AG8001-0203 AG8001-0202 CAAG8001-0202 CAAG8004-0202 CAAG8001-0201 CAAG8004-0204 CA8003-0205 CAAG8004-0203 AG8003-0205 AG8003-0204 AG8003-0203 AG8003-0202 AG8004-0201 AG8004-0202 CAAG8003-0203 CAAG8003-0202 CAAG8003-0204 CAAG8002-0203 CAAG8002-0204 CAAG8002-0201 CAAG8010-0401 AG8002-0203 AG8002-0204 AG8002-0201 AG8010-0401 CA8004-0201
Liner O-Rings for Third Party GC Instruments, Agilent Technologies
Seals Sealing Rings
Agilent provides a range of high-quality liner O-rings for various third party GC systems.
  • Liners are sealed in the inlet with fluor elastomer or graphite O-rings
  • Graphite O-rings are used when inlet temperatures exceed 350 °C
  • Fluor elastomer O-rings are easier to replace than graphite O-rings, which deteriorate more readily under certain conditions
Liner O-rings are used to form a proper seal around the outside of the inlet liner between the inlet weldment and the turn-top. O-rings are composed of either graphite or fluorocarbon material, Graphite O-rings are used when operating inlet temperatures exceed 350°C. Agilent fluor elastomer or graphite liner O-rings for third-party GC systems help to effectively seal the inlet liner. Fluor elastomer O-rings are provided ready for use and feature a proprietary two-step cleaning and conditioning process. This process eliminates outgassing of contaminants. The plasma treatment results in a nonstick, contaminant-free surface. A novel dial package also conveniently delivers one clean O-ring at a time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty