Would you like to log in again?
Standard Racks and Vessels, Agilent Technologies
Supplier: Agilent Technologies
810177600 810177600
AG810177600EA 9.95 USD InStock
CAAG810177600 AG810177600
Standard Racks and Vessels, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Standard racks and compatible vessels for SPS 4 autosampler used with AAS, ICP-OES, MP-AES, and ICP-MS.
  • Choose from 2 standard rack options available for the SPS 4 autosampler
  • Higher throughput with 34-well (12 wells for 29 mm ext.Ø tubes + 22 wells for 17 mm ext.Ø tubes) - not supported with the Mass Hunter software for ICP-MS
  • Compatible with larger volume sample vessels in 5 well (for 60 mm ext.Ø bottles) - not supported with the SpectrAA software for AAS
The Agilent SPS 4 autosampler is designed to automate the presentation of liquid samples. The samples are drawn from tubes held in racks loaded on the rack location mat , and presented to the instrument for analysis. The Agilent SPS 4 can accommodate a variety of fixtures, including a standards rack, four sample racks, and other accessories.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty