Would you like to log in again?
ZORBAX® Bio-SCX Series II Columns, Agilent Technologies
Supplier: Agilent Technologies
ZORBAX®
820950-903 820950-903
AG820950-903PK 286 USD InStock
AG820950-903 CA820950-903
ZORBAX® Bio-SCX Series II Columns, Agilent Technologies
Chromatography Columns
ZORBAX® Bio-SCX Series II Columns are designed for optimized 2D separations of peptides and proteins using LC-MS.
  • Offered in 0.3 Int.Ø×35L mm and 0.8 Int.Ø×50L mm column configurations
  • pH range of 2.5 to 8.5
This packing is based on ultrapure 3.5 µm ZORBAX® silica particles, bonded with a biofriendly polymer that is functionalized with sulfonic acid groups. This gives strong retention and good peak shape in the ion exchange step of 2D-analysis of peptides and proteins. View our full range of BioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty