Would you like to log in again?
InfinityLab Poroshell 120 HILIC, Agilent Technologies
Supplier: Agilent Technologies
Poroshell
683975-324 683975-601 689975-924 685775-601 683970-924 689975-324 679775-924 823750-948 685975-924 689675-924 821725-947 679775-924 673775-924 820750-933 673775-924 685975-324 689975-601 689975-601 689775-924 683675-924 680970-924 685975-924 685675-924 821725-947 685975-324 689975-924 689675-924 689775-601 685775-601 683975-324 683775-924 685675-924 689775-924 683975-924 680970-924 683675-924 689775-601 683970-924 683975-924 683975-601 820750-933 683775-924 823750-948 685775-924 689975-324 685775-924 675775-924 675775-924
AG683675-924EA 1840 USD InStock
CAAG680970-924 CAAG675775-924 CAAG683775-924 CAAG685775-924 CAAG673775-924 CAAG820750-933 CAAG685775-601 CAAG679775-924 CA821725-947 AG683675-924 AG689675-924 AG685675-924 AG820750-933 CAAG689975-601 AG821725-947 AG689775-924 AG685975-924 CAAG689975-924 AG683975-924 CAAG683675-924 CAAG689975-324 AG683970-924 AG685975-324 CAAG823750-948 AG689975-924 CAAG685675-924 AG683975-324 AG683975-601 AG689975-601 CAAG689775-601 AG673775-924 CAAG683975-924 CAAG683970-924 AG689975-324 CAAG689675-924 CAAG689775-924 AG683775-924 AG680970-924 AG685775-924 CAAG683975-324 CAAG685975-324 AG823750-948 AG685775-601 CAAG683975-601 CAAG685975-924 AG689775-601 AG679775-924 AG675775-924
InfinityLab Poroshell 120 HILIC, Agilent Technologies
Chromatography Columns
Superficially porous particles for high-resolution HILIC separations with short equilibration times.
  • Superficially porous, 'core-shell' particles that provide improved throughput, shorter equilibration times, and higher resolution
  • A scalable family of particles—1.9, 2.7, and 4 μm—enable you to get the best from your methods and instruments
  • Separate many polar compound classes by choosing from orthogonal bare silica (HILIC), poly-hydroxy fructan (HILIC-OH5), or zwitterionic (HILIC-Z) phases
  • Fully compatible with Agilent InfinityLab deactivator additive, which mproves sensitivity in challenging analyses of phosphorylated and carboxylated compounds with stainless steel LC/MS systems
  • Part of the InfinityLab family - designed for ease-of-use and maximum compatibility with Agilent instruments and supplies
  • Long column life with uniquely robust 1.9 μm low-dispersion UHPLC columns
  • Reduced need for sample preparation due to 2 μm frits on 2.7 μm and 4 μm columns
  • UHPLC guard columns to further extend the life of your analytical column
  • Enhanced usability, traceability, and security with a preprogrammed ID tag
Agilent InfinityLab Poroshell 120 HILIC columns for hydrophilic interaction liquid chromatography (HILIC) are designed for the retention and high-resolution separation of small polar analytes. The superficially porous character of the particles decreases equilibration times to improve throughput for both isocratic and gradient separations. Agilent offers three unique, orthogonal HILIC phases: bare silica (HILIC), polyhydroxy fructan (HILIC-OH5), and zwitterionic (HILIC-Z). The robustness and reproducibility of HILIC separations on Poroshell columns help to switch from challenging, often MS-incompatible ion pair chromatography and normal phase applications.
Agilent InfinityLab Poroshell 120 HILIC columns for hydrophilic interaction liquid chromatography (HILIC) are designed for the retention and high-resolution separation of small polar analytes. The superficially porous character of the particles decreases equilibration times to improve throughput for both isocratic and gradient separations. Agilent offers three unique, orthogonal HILIC phases: bare silica (HILIC), polyhydroxy fructan (HILIC-OH5), and zwitterionic (HILIC-Z).
The robustness and reproducibility of HILIC separations on Poroshell columns help to switch from challenging, often MS-incompatible ion pair chromatography and normal phase applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty