Would you like to log in again?
AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Supplier: Agilent Technologies
695975-322 695975-322 823750-946 823750-946
AG823750-946PK 882 USD InStock
CAAG695975-322 AG823750-946 AG695975-322 CAAG823750-946
AdvanceBio Amino Acid Analysis (AAA), Agilent Technologies
Chromatography Columns
For fast and reproducible pre-column derivatization and separation of amino acids in protein hydrolysates and cell culture media.
  • Reliable results - high-resolution separations delivered by efficient Poroshell particle morphology
  • Reduced costs - long column lifetimes from robust, high-pH resistant, chemically modified silica
  • Increased flexibility - compatibile with both HPLC and UHPLC systems via 2.7 μm diameter particles
  • Quality control - AdvanceBio AAA columns are batch tested with amino acid standards to ensure quality
  • Easy ordering - standards and reagents available as kits
  • Automated online derivatization - with Agilent analytical injection systems
AdvanceBio Amino Acid Analysis (AAA) includes proven reagents for amino acid derivatization, a ready-to-use amino acid standards kit, columns based on innovative Poroshell technology, and expert support.
AdvanceBio AAA delivers fast, sensitive, and reproducible separations of amino acids in biological samples, protein hydrolysates, and cell culture media. Used alongside Agilent InfinityLab LC Series instruments, AdvanceBio AAA provides a complete solution for amino acid analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty