Would you like to log in again?
ZORBAX for NP/SFC, Agilent Technologies
Supplier: Agilent
959961-901 959758-901 820950-908 828700-901 880952-701 820668-931 959961-901 828975-901 959759-901 880952-201 860700-708 820668-901 828975-901 880952-201 821125-919 820675-111 959758-901 880952-205 959743-901 820675-111 820950-901 820950-908 880952-205 821725-938 880952-701 880952-208 820668-931 880953-901 820950-901 820950-919 820950-919 827975-901 821125-919 827975-301 827975-901 828700-901 827975-301 883952-701 959743-901 821725-938 820668-901 860700-708 959793-901 959793-901 883952-701 880953-901 959759-901 880952-208
AG821725-938PK 713.33 USD InStock
AG821725-938 AG820668-901 AG820950-919 AG860700-708 CAAG828700-901 AG959743-901 CA820950-901 CAAG860700-708 AG820675-111 CAAG821725-938 CAAG820668-901 CA820950-908 AG880953-901 CAAG959961-901 CAAG959758-901 AG828700-901 CAAG827975-301 AG880952-201 CA820675-111 AG959759-901 AG820668-931 AG821125-919 AG883952-701 AG880952-701 AG820950-908 AG880952-205 CAAG959793-901 AG959793-901 AG880952-208 CAAG880953-901 AG959961-901 AG820950-901 CAAG828975-901 CAAG959759-901 CAAG827975-901 CAAG820668-931 CA880952-201 CAAG880952-205 AG959758-901 AG827975-301 CAAG880952-208 CA880952-701 CA820950-919 CA821125-919 CAAG883952-701 AG827975-901 AG828975-901 CAAG959743-901
ZORBAX for NP/SFC, Agilent Technologies
Chromatography Columns
Fully porous columns based on ZORBAX silica for normal phase LC and SFC separations.
  • Traditional fully porous ZORBAX silica particles available with different stationary phases and particle sizes
  • Master challenging normal phase applications with orthogonal chemistries such as ZORBAX Rx-Sil, CN, and NH2 phases
  • Discover the robust, general-purpose ZORBAX HILIC Plus and Rx-Sil columns for SFC, available in 1.8 µm and 5 µm particle sizes
  • Use Agilent Application Finder to find application notes of ZORBAX columns used in NP-LC, SFC, and SFC/MS
Agilent ZORBAX fully porous particle columns for normal phase HPLC and supercritical fluid chromatography (SFC) are designed for the retention and separation of lipophilic compounds. The unique ZORBAX chemistries deliver outstanding efficiency in both achiral SFC and normal phase chromatography due to the similar retention mechanisms of both LC modes on the polar stationary phases used. The recommended ZORBAX phases are Rx-Sil, amino (NH2), cyano (CN), Eclipse XDB-CN, and HILIC Plus (HILIC phases in SFC only).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty