A range of high-quality supplies for LC/MSD to ensure continued high levels of system performance.
- Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
- Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent provides a range of high-quality supplies for LC/MSD to ensure continued high levels of system performance. These supplies include common parts such as ion injectors, capillaries, electron multipliers, and nebulizer needles. LC/MS source supplies ensure maximum source longevity, consistency, and integrity.
Agilent also provides gas purifiers including moisture, hydrocarbon, and universal traps, and a range of specialist tools suitable for LC/MSD calibration, operation, and maintenance. All components and supplies are precision-engineered to Agilent's exacting quality standards.