Would you like to log in again?
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
9301-6525 5063-6531 5065-4421 5063-6531 9301-6530 9301-6524 9301-6529 5043-1339 5067-6871 9301-6342 9301-6525 9301-6341 9301-6532 5043-1196 9301-6526 5065-4421 5043-1986 9301-6530 9301-6526 9301-6523 5043-1221 5043-1339 5043-1337 5067-6871 5043-1986 9301-6528 9301-6528 9301-6529 9301-6342 5043-1816 9301-6531 G4220-68070 G4220-68070 5043-1340 9301-6341 9301-6531 5043-1337 9301-6527 9301-6532 5043-1340 5043-1816 5043-1196 9301-6524 9301-6527 9301-6523 5043-1221
AG5043-1196EA 39.21 USD InStock
CAAG9301-6530 CAAG5043-1337 CAAG9301-6531 CAAG5043-1339 CA5065-4421 AG5043-1196 CAAG9301-6532 CA76514-668 AG5065-4421 AG9301-6530 AG9301-6531 AG9301-6532 CAAG5043-1816 CA5043-1221 AGG4220-68070 AG5067-6871 AG5043-1816 CAAG5043-1196 AG5043-1339 AG5043-1337 CAAG5067-6871 CAAG5043-1986 CA9301-6524 CA9301-6523 CA9301-6526 AG9301-6341 AG5043-1221 AG9301-6342 CA9301-6525 CAAGG422068070 AG5043-1340 AG9301-6523 AG9301-6524 AG9301-6525 AG9301-6526 CA9301-6528 CA9301-6527 AG5063-6531 AG9301-6527 AG9301-6528 AG9301-6529 CAAG5043-1340 CA9301-6342 CA9301-6341 AG5043-1986 CAAG9301-6529
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Designed to provide high levels of performance for your LC applications.
  • Slim profile and ergonomically shaped gripping zones on both sides of the bottle to promote easier and safer use
  • A predefined labeling area with self-adhesive labels and colorful silicone bottle tags let you clearly and unambiguously identify solvent compositions
  • An easy-to-read durable white ceramic print layout clearly indicates the nominal volume of the bottle and allows you to quickly determine the contents of the bottle
  • Bottles have a GL 45 thread and are compatible with Agilent Stay Safe caps
InfinityLab solvent glass reservoirs are designed to provide high levels of performance for your LC applications. The 1 liter and 2 liter reservoirs are available in amber and in clear, for situations where sample visibility is important or to avoid algae growth. All reservoirs can be used with the InfinityLab Stay Safe caps. Agilent solvent bottles have been redesigned for easier use and safer handling, lowering the risk of accidents in the lab. These redesigned solvent bottles are compatible with your InfinityLab LC Series instruments, and can be incorporated seamlessly into your LC workflow.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty