Would you like to log in again?
Capillary Tubing and Nebulizer Maintenance Supplies, Agilent Technologies
Supplier: Agilent Technologies
9910044000 9910024800 9910024800 9910044000
AG9910044000PK 43.1 USD InStock
AG9910044000 CA9910044000 AG9910024800 CA9910024800
Capillary Tubing and Nebulizer Maintenance Supplies, Agilent Technologies
Tubing Chromatography Tubing
Narrow bore and wide bore capillary tubing for various applications, solvents, and sample types.
  • Choose the type of capillary tubing to optimize the performance required for your application with your Agilent nebulizer
  • Use narrow bore capillary tubing (0.38 mm Int.Ø) for aqueous solutions and samples that have relatively low levels of dissolved solids
  • Use wide bore capillary tubing (0.64 mm Int.Ø) for samples that have high levels of dissolved salts and organic solvents
The performance of Agilent nebulizers can be adjusted by switching the type of capillary tubing fitted to the nebulizer.;Precut lengths of narrow bore capillary tubing (0.38 mm Int.Ø) with a flared end is recommended for use with barrel nebulizers in Hi-vac mode and with aqueous solutions. This tubing is also recommended for samples that have relatively low levels of dissolved solids
Wide bore capillary tubing (0.64 mm Int.Ø) is recommended for use with barrel nebulizers in the high solids position and with samples that have high levels of dissolved salts. This tubing is also recommended for use with the adjustable nebulizer when aspirating organic solvents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty