Would you like to log in again?
ACT 80 Supplies, Agilent Technologies
Supplier: Agilent Technologies
9910054400 9910054400
AG9910054400PK 1206 USD InStock
CAAG9910054400 AG9910054400
ACT 80 Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Typical supplies forAgilent ACT 80 Atom Concentrator Tube used with Agilent flame AA Spectrometers.
  • Simple, cost effective and offers benefits in low level analyses
  • Increases sensitivity by 2 to 3 times for elements determined using the air/acetylene flame
  • Improves detection limits by up to 3 fold
  • Measurement is as fast as normal flame determinations, and can be used with manual or automated sampling
  • Suitable for use with the air-acetylene burner and compatible with any Agilent flame AA instrument
The ACT-80 Atom Concentrator Tube is a quartz tube that is positioned in the air-acetylene flame to enhance the sensitivity of flame AA by 2 or 3 times when compared to that of normal flame AA. An improvement in detection limit is also achieved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty