Would you like to log in again?
CrossLab Internal Standard Kits and Supplies for ICP-OES, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
9910124000 9910124100 1610132400 9910124000 9910124100 1610132400
AG1610132400EA 16.15 USD InStock
AG1610132400 CA1610132400 AG9910124000 AG9910124100 CAAG9910124100 CAAG9910124000
CrossLab Internal Standard Kits and Supplies for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Internal standard kit for ICP-OES. Use with organic samples.
  • Extend quality and durability of instruments;Reduce sample prep time with on-line addition of internal standard;Save up to 15% compared to purchasing supplies individualy
The ICP-OES internal standard kit includes standard PVC pump tubing, EzyFit connectors, five Y pieces and connecting tubing. Use it with aqueous, organic, or high TDS samples to meet your toughest measurement challenges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty