Would you like to log in again?
Polaris, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A2007050X020 A2000MG A2001150C046 A2020250X046 A2021100X020 A2000050X020 A2000MG2 A2000250X100 A2000250X100 A2011150X046 A2001050X030 A2001150X020 A2001150X030 A2000MG2 A2007050X020 A2007100X046 A2001100X020 A2001050X030 A2010250X046 A2006250X046 A2010250X046 A2000050X020 A2000MG A2021250X046 A2001150X020 A2001250X046 A2021100X020 A2000250X020 A2000050X046 A2021250X046 A2006250X046 A2001100X020 A2011150X046 A2001250X046 A2000250X020 A2000100X046 A2020250X046 A2001150C046 A2000050X046 A2000100X046 A2007100X046 A2001150X030
AGA2000050X020EA 621 USD InStock
AGA2000050X020 CAAGA2000MG CAG2001050X030 AGA2000050X046 CAAGA2000MG2 AGA2007100X046 AGA2001150X030 CAG2001150C046 AGA2001100X020 AGA2020250X046 CAG2007050X020 AGA2007050X020 AGA2021250X046 AGA2006250X046 CAG2000250X020 AGA2000100X046 CAG2006250X046 CAAGA220250X46 AGA2010250X046 CAG2001250X046 CAG2000250X100 AGA2000250X100 CAG2001150X030 AGA2001250X046 CAG2007100X046 CAG2000100X046 AGA2000250X020 AGA2021100X020 CAG2000050X020 CAG2021100X020 CAAGA201150X20 CAG2001100X020 CAG2000050X046 AGA2000MG AGA2001150X020 AGA2001150C046 CAG2010250X046 AGA2001050X030 CAG2021250X046 AGA2000MG2 CAG2011150X046 AGA2011150X046
Polaris, Agilent Technologies
Chromatography Columns
Polaris HPLC columns offer superior retention of polar compounds in 100% water.
  • Separate mixtures of polar and nonpolar analytes reliably and with excellent peak shape
  • Polar-embedded C8 and C18 bonded phases are compatible with 100% water
  • Wide range of alternative selectivities that comply with USP L1 and USP L7
Agilent Polaris HPLC columns offer many polar phase chemistries designed separate mixtures of polar and nonpolar analytes reliably and with excellent peak shapes. The polar-embedded C8 and C18 bonded phases are compatible with 100% water, and offer alternative selectivities while still complying with USP L1 and USP L7. The combination of high carbon load, ultrapure silica, and silanol shielding provides superior peak shape and retention with highly polar samples.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty