Would you like to log in again?
Bond Elut Plexa Plates, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
A4969030HF A3969010 A3969010 A3969030B A3969030B A4969030HF A4969010 A4969010
AGA4969030HFEA 431 USD InStock
AGA4969030HF CAAGA4969010 AGA4969010 AGA3969030B AGA3969010 CAAGA3969010 CAAGA4969030HF CAAGA3969030B
Bond Elut Plexa Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Plexa offers simple extraction, minimal method development, and improved cleanliness.
  • Nonpolar retention mechanism
  • Improved extract cleanliness minimizes sample matrix interferences
  • Simple methods are amenable to a very broad range of analytes
  • Fast flow, reproducible performance, and ease-of-use
Bond Elut Plexa offers easy-to-use methods for general purpose extraction. A unique polymeric architecture with a nonretentive, hydroxylated, amide-free surface, and a nonpolar PS-DVB core retains small molecules. Binding of proteins and lipids on the polymer surface is minimized, giving cleaner samples and reduced ion suppression.
Bond Elut Plexa offers easy-to-use methods for general purpose extraction. A unique polymeric architecture with a nonretentive, hydroxylated, amide-free surface, and a nonpolar PS-DVB core retains small molecules. Binding of proteins and lipids on the polymer surface is minimized, giving cleaner samples and reduced ion suppression.
Plexa is ideal for high-throughput assays requiring validated performance with minimal method development. The standard nonpolar retention mechanism is applicable to almost any analyte type. Plexa Polymeric SPE is also available in ion exchange SPE formats, Plexa PCX SPE for cation exchange, and Plexa PAX for anion exchange.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty