Would you like to log in again?
CrossLab Bond Elut Focus, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
A5106010 A5306022 A5306022 A5106010
AGA5106010PK 333 USD InStock
AGA5106010 CAAGA5106010 CAAGA5306022 AGA5306022
CrossLab Bond Elut Focus, Agilent Technologies
Chromatography Columns
Bond Elut Focus delivers outstanding retention for polar and nonpolar analytes.
  • Simple, universal method reduces method validation times
  • 'Power rinse' delivers cleaner extracts, improving sensitivity
  • Hydrogen bond-donor capability increases retention of basic analytes
Bond Elut Focus features a unique amide-functionalized, polar-enhanced sorbent technology delivering outstanding retention. The extraction method eliminates the complex pH adjustments necessary with ion exchange and mixed mode methods. The unique ligand chemistry on the sorbent dramatically increases the retention of basic analytes.
The improved retention for polar analytes with Focus allows the use of a strong 'power rinse' without causing analyte loss. Rigorous washes using 10 to 20% MeOH can be used to aggressively remove endogenous matrix interferences. For very polar analytes that are held by strictly polar interactions, washes of up to 100% can be used.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty