Would you like to log in again?
Captiva ND and ND Lipids Plates, Agilent Technologies
Supplier: Agilent
Captiva
A5969002 A59640002I A59640002V A5969002 A59640002I A59640002RK A59640002B A59640002SK A5969045 A5969045
AGA5969045PK 535.76 USD InStock
CAA5969045 AGA5969045 101473-998 101404-466 101473-996 AGA59640002I 101473-994 CAAGA59640002I AGA5969002 CAA5969002
Captiva ND and ND Lipids Plates, Agilent Technologies
Microplates Filter Plates
Captiva ND enables in-well protein precipitation; Captiva ND Lipids removes biofluid phospholipids.
  • Fully automatable protein precipitation
  • Reduced costs and increased productivity due to less instrument downtime
  • Simple and easy-to-use nondrip protein precipitation plate
  • Resists organic solvent flow until vacuum is applied, allowing for streamlined, fully automatable workflow
  • Dual depth filter design allows for fast uniform treatment of samples while avoiding membrane plugging or lost samples
  • In-well filter design removes proteins as efficiently as centrifugation while avoiding additional handling steps and accidental pellet transfer
Captiva ND cartridges and plates are a simple-to-use filtration device designed for high-throughput, automated, in-well protein precipitation. With a unique nondrip (ND) membrane, Captiva ND products enable solvent-first protein precipitation using methanol or acetonitrile. The dual-filter design delivers fast, uniform flow, avoiding filter plugging and ensuring reliable filtrate recovery.
In addition, Captiva ND Lipids cartridges and plates are designed to effectively remove phospholipids from biofluids. The removal of lipid compounds results in enhanced sensitivity, better peak shapes, and more reproducible retention times. Captiva ND and ND Lipids products ensure high sample throughput and help to reduce instrumentation downtime in the laboratory.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty