The Pursuit family of HPLC columns offer many chemistries with easy scale up to preparative HPLC.
- Available in many dimensions, including preparative HPLC columns
- Includes both traditional (Pursuit) and high carbon load (Pursuit XRS) bonding
The Agilent Pursuit family of HPLC columns offers reliable performance in various common particle sizes and phase chemistries, including USP L1, USP L7, and USP L11. Pursuit columns are available in many dimensions, including preparative HPLC columns. In addition, both traditional (Pursuit) and high carbon load (Pursuit XRS) bonding are available.