Would you like to log in again?
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5022-6503 01018-67305 G2258-87315 G4280-60034 5062-2463 RF0110T-7 G4280-60034 5042-6463 RF0110T-3 CP4105 5042-6461 R0PTF20KIT G2258-87300 RF0111T-2 G1375-87326 5062-2463 5042-6462 RF0111T-2 0890-1763 5043-1540 0890-1762 G2258-87300 G2258-87312 R0PTF20KIT AL35720 RF0112T-16 G1313-87300 AL35721 5022-6503 G1364-86708 G2258-87316 G2258-87310 0890-1915 RF0112T-6 RF0110T-7 RF0110T-3 RF0110T-60 0890-1761 CP4105 G2258-87310 AL35724 G1311-67304 G2258-87316 0890-1761 G1375-87326 RF0112T-50 01090-87305 G2258-87314 RF0111T-8 5062-2461 G1364-86708 01090-87305 RF0112T-6 5042-6461 5185-5817 5500-1228 5043-1540 5185-5817 5062-2461 G2258-87313 RF0103T G2258-87315 RF0112T-50 AL35724 G1311-67304 0890-1915 5067-4680 UC1535 UC1535 5500-1228 AL35721 0890-1763 RF0112T-13 RF0112T-16 5067-4680 01018-67305 G4280-67304 G2258-87312 RF0103T 5042-6463 RF0110T-60 G2258-87314 G4280-67304 0890-1762 G2258-87313 RF0111T-8 AL35720 RF0112T-13 G1313-87300 5042-6462
AGAL35720EA 155 USD InStock
AGAL35720 CAAGRF0110T-7 AGAL35721 AGAL35724 CAAGRF0110T-3 CAAL35720 CAAG5042-6463 CA5062-2461 CAAGG428067304 AGCP4105 CA5062-2463 CAAGG428060034 AG5500-1228 CA5022-6503 AG0890-1915 CAAGRF0103T CAG5042-6462 CAAGRF0110T-60 CAAGRF0111T-8 CAAGAL35721 CAAGRF0111T-2 AGG4280-60034 AG0890-1763 CAAGG225887313 AG0890-1762 CAAGAL35724 CAAGG225887314 AGG4280-67304 CAAGG225887315 AG0890-1761 CAAGRF0112T-50 CAAGG225887316 CAAGR0PTF20KIT CAAG5067-4680 CAAGRF0112T-13 CAAGG225887310 CAAGG137587326 CAAGG225887312 AGRF0103T CAAG5043-1540 CAAGRF0112T-16 AG5022-6503 CAG1311-67304 CAG1313-87300 AGRF0112T-6 CA5042-6461 AG5185-5817 AGG1311-67304 CAAG5185-5817 CAAGRF0112T-6 AGG1375-87326 CAAGCP4105 AG5062-2463 CA76514-524 AG5062-2461 CA76514-526 AGG2258-87310 AGG2258-87313 AGG2258-87312 AGG2258-87315 AGG2258-87314 AGG2258-87316 CAAGG225887300 AGG1364-86708 CAAGG136486708 AG01090-87305 CA01018-67305 AGRF0111T-2 AG5043-1540 AGRF0112T-50 AGUC1535 AGRF0111T-8 AGRF0112T-13 CA0890-1761 CA01090-87305 CA0890-1762 AGRF0112T-16 AGG2258-87300 CAAGUC1535 AGR0PTF20KIT CAAG5500-1228 AGG1313-87300 AG5042-6462 AG5067-4680 AG5042-6463 AGRF0110T-60 AGRF0110T-7 AG5042-6461 AG01018-67305 AGRF0110T-3
CrossLab Tubing for HPLC, Agilent Technologies
Tubing Chromatography Tubing
A range of high-quality tubing made from PEEK, PTFE, and other materials.
  • PEEK tubing with ¹/₁₆" (1.6 mm) OD that are flexible and easy to cut to desired lengths, as well as color-coded for easy tracking
  • Tubing made of PTFE, PP, or FEP are available for special conditions
  • Available accessories include tubing cutters, ferrules, unions, adapters, and more
Agilent provides a range of high-quality tubing made from PEEK, PTFE, and other materials to ensure certain connnections in your LC system. All tubings are manufactured to the highest quality standards to protect the integrity of your results at every step of your LC flow path.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty