Would you like to log in again?
CrossLab Thermal Desorption Supplies for GC, Agilent Technologies
Supplier: Agilent
CrossLab
MKI-U-T1KORI MKI-U-T18325-2S C-TBME10 C-TBP1C1C C-TBP1TC 5182-0946 MKI-UTE-5099 C-AT010C MKI-U-T15ATA-2S C-CF020 MKI-UTD-5064 MKI-C-CPLOK MKI-U-T12ME-2S C-PL010 C-PL010 MKI-MTD-1204 MKI-U-T17O3P-2S MKI-UTD-5105 MKI-U-T11GPC-2S MKI-UTD-5105 MKI-U-T17O3P-2S MKI-UTD-5064 C-TBE10 C-GT010 MKI-U-T1KORI C-DF010 C-UN010C C-GAT010C 5182-0946 C-TBME10 C-TBE10 C-AT010C MKI-U-T18325-2S MKI-C-CPLOK MKI-U-T3ATX-2S MKI-U-T12ME-2S MKI-SERUTD-5065 MKI-UTE-5099 MKI-U-T15ATA-2S C-DF010 C-GT010 MKI-U-T3ATX-2S MKI-U-T11GPC-2S MKI-MTD-1169 MKI-U-T13DHS-2S C-UN010C MKI-MTD-1204 C-CF020 MKI-SERUTD-5065 C-TBP1C1C C-GAT010C MKI-MTD-1169 C-TBP1TC MKI-U-T13DHS-2S C-HY010C C-HY010C
AGC-TBP1C1CPK 2214.81 USD InStock
AGC-TBP1C1C CAAGC-TBP1C1C CAAGMKIUTD5064 AGMKI-UTE-5099 CAAGMKIMTD1204 CAAGMKIMTD1169 AGC-PL010 CAAGMKIUTD5105 AGMKI-MTD-1204 AGMKI-MTD-1169 AGC-CF020 CAAGMKIUT1KORI AGMKI-C-CPLOK AGC-TBE10 CAGSERUTD5065 CAGMKIUT12ME2S AGC-DF010 CAGMKIUT13DHS2 AGC-UN010C AGMKI-U-T11GPC CAGMKIUT3ATX2S CAAGC-CF020 CAAGC-PL010 AGC-TBME10 CAGMKIUT17O3P2 AGC-HY010C AGMKI-U-T13DHS AGC-GT010 CAGMKIUT11GPC2 CAAGC-UN010C AGMKI-U-T1KORI CAAGC-DF010 CAAGC-HY010C CAAG5182-0946 CAAGC-TBME10 AG5182-0946 AGC-TBP1TC AGMKI-UTD-5105 CAAGC-GAT010C AGMKI-UTD-5064 CAAGMKIUTE5099 CAAGC-GT010 AGMKI-U-T15ATA AGC-AT010C AGMKI-SERUTD CAGMKIUT183252 AGMKI-U-T3ATX CAAGMKICCPLOK CAAGC-AT010C AGMKI-U-T12ME CAGMKIUT15ATA2 AGMKI-U-T18325 CAAGC-TBE10 AGC-GAT010C CAAGC-TBP1TC AGMKI-U-T17O3P
CrossLab Thermal Desorption Supplies for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent offers the supplies you need for your Markes Thermal Desorber.
  • Quantitative sample recollection of all the split flows enables repeat analysis of critical samples, easy method validation, and overcomes the one-shot limitation of conventional TD systems
  • Electronic tube tagging (TubeTAG): RFID tube tags are available for industry standard sorbent tubes
  • Diffusion locking (DiffLok) for enhanced sample integrity and robust (mechanically simple) automation
  • Patented inert valving for compatibility with every TD application on a single analytical platform - ultravolatiles, semivolatiles (up to n-C40), plus reactive species - mercaptans, CS gas, and so forth - all on one TD system
  • Automated internal standard introduction onto blank as well as sampled tubes
  • Electronic pneumatics control (EPC) of carrier gas through the thermal desorber ensures consistent compound retention time independent of split flow
  • Electrically cooled sorbent trapping with uniquely fast trap heating rates for splitless capillary GC operation and optimum sensitivity without risk of ice formation
  • Offline conditioning for multiple tubes without the need to blank-off unused tube connections
  • Specialist sorbent tubes: Certified reference standards, SafeLok tubes, Silcosteel tubes
The analytical technique known as thermal desorption (TD) provides the ability to measure volatile contaminants in air, toys, building materials, car cabins, and food. When the TD systems from Markes International are combined with Agilent GC and GC/MS systems, the results meet current requirements with the sensitivity and dynamic range to meet future needs.
Markes International TD platforms allow analysis of single tubes, real-time air samples, and canisters with options for automated analysis. Markes has introduced several key technical advances that now set the standard for TD instrumentation. Agilent offers the supplies you need for your Markes Thermal Desorber.
A range of unique sampling tools for measuring volatile and semivolatile organics in challenging matrices: such as liquids/solids/emulsions, in-situ soil, polymers, natural products, and construction products.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty