Would you like to log in again?
CrossLab Tubing, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CP4056 1300502506 CP4260 CP4008 5190-6983 CP4032 CP4024 CP4026 CP4006 CP4005 5062-2419 1300502506 7157-0210 CP4049 CP4025 CP4003 AL3002 CP4025 AL3000 AL3003 5180-4196 CP4013 5180-4196 R000383005 CP4006 CP4026 CP4032 7157-0210 CP4024 CP4008 CP4056 5021-7107 5062-2419 AL3003 CP4260 5021-7107 AL3000 5190-6983 R000383005 CP4013 CP4003 AL3002 CP4049 CP4005
AG5190-6983EA 348 USD InStock
CAAGCP4008 AG5190-6983 AG5180-4196 CAAGCP4025 CAAGCP4003 CAAGCP4026 CAAGCP4005 CAAGCP4049 CAAGCP4006 AGCP4026 AGAL3002 CA76514-722 AGCP4003 AGCP4025 AGAL3003 AGCP4024 AGCP4008 CAAG5190-6983 AGCP4006 AGCP4049 AGCP4005 AGCP4260 AGAL3000 CA5021-7107 CAAGCP4024 CAAGCP4260 CAAGAL3000 CAAGAL3003 CAAGAL3002 CAAGR000383005 AG1300502506 AGCP4013 AGCP4056 AGR000383005 CAAG5062-2419 AG5021-7107 CA1300502506 AGCP4032 CAAG7157-0210 CAAGCP4032 CAAGCP4056 CAAGCP4013 AG5062-2419 AG7157-0210
CrossLab Tubing, Agilent Technologies
Tubing Chromatography Tubing
Copper, stainless steel, and UltiMetal Plus tubing for GC applications.
  • Copper tubing can be used for most applications, except those requiring the highest purity gas delivery system
  • Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
Agilent GC tubing includes high-quality copper, stainless steel, and UltiMetal Plus tubing, which are manufactured and cleaned for chromatographic use. Agilent tubing comes in different lengths and depending on your flow requirements, is available in ¹/₁₆, ¹/₈, or ¹/₄". UltiMetal Plus tubing is available in ¹/₁₆" for inert environments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty