Would you like to log in again?
CP-Sil 5 CB Columns, Agilent Technologies
Supplier: Agilent
CP7710 CP8690 CP7625 CP7693 CP7475 CP7710 CP8870 CP7760 CP8674 CP7625 CP7529 CP8688 CP7475 CP7680 CP8770 CP7529 CP7170 CP7709 CP8688I5 CP7692 CP7135 CP7690 CP7640 CP8688I5 CP8735 CP7684 CP7670 CP8741 CP7750 CP7693 CP8770 CP6666 CP7709 CP6670 CP8690 CP8674 CP8688 CP7682 CP7682 CP8677 CP6670 CP7692 CP7170 CP7640 CP7687 CP7739 CP7140 CP7675 CP7750 CP8780 CP7675 CP7680 CP8741 CP6666 CP7135 CP8735 CP7760 CP8870 CP7685 CP8677 CP8780 CP7690 CP7670 CP7687 CP7685 CP7684 CP7140 CP7739
AGCP7739EA 687.65 USD InStock
CAAGCP6666 CAAGCP7710 AGCP7739 CAAGCP8688I5 AGCP7693 AGCP8741 AGCP7135 AGCP7690 AGCP7692 AGCP8780 CAAGCP7475 CAAGCP7750 AGCP7170 CAAGCP8688 CAAGCP7675 CAAGCP7670 AGCP7709 CAAGCP7625 AGCP7625 AGCP8677 CAAGCP7709 AGCP8870 CAAGCP7140 AGCP8674 AGCP7140 CAAGCP8674 CAAGCP8677 CAAGCP8870 CAAGCP7739 AGCP8688 AGCP7675 AGCP7710 AGCP6666 CAAGCP7170 AGCP7475 CAAGCP7690 AGCP7750 AGCP7670 CAAGCP8741 CAAGCP7135 CAAGCP7692 CAAGCP8780 CAAGCP7693 CACP7684 CAAGCP7529 AGCP7529 CAAGCP8735 AGCP8735 AGCP7687 AGCP7682 AGCP6670 AGCP7760 AGCP7640 AGCP7684 CAAGCP8690 AGCP7685 AGCP8690 AGCP7680 AGCP8770 CAAGCP7685 AGCP8688I5 CAAGCP7640 CAAGCP7687 CAAGCP8770 CAAGCP7680 CAAGCP7760 CAAGCP6670 CAAGCP7682
CP-Sil 5 CB Columns, Agilent Technologies
Chromatography Columns
A low-polarity, general-purpose, 100% dimethylpolysiloxane (PDMS) column ideal for a wide range of applications.
  • 100% dimethylpolysiloxane (PDMS)
  • Nonpolar
  • General purpose phase
  • Bonded, crosslinked, and solvent rinsible
  • Separates almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range
  • Available in Fused Silica or UltiMetal
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, ZB-1
Agilent J&W CP-Sil 5 CB is a general-purpose, nonpolar column containing a 100% dimethylpolysiloxane (PDMS) phase. Separation is almost entirely based on boiling points, making this column suitable for a wide range of applications with a broad temperature range.
Due to extensive crosslinking, CP-Sil 5 CB is highly inert and withstands large solvent injections, guaranteeing reproducibility and ensuring maximum column lifetime. Available in UltiMetal for the highest operating temperatures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty