Would you like to log in again?
J&W Porabond PoraBOND Q Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CP7350 CP7354 CP7352 CP7351 CP7353 CP7351 CP7353I5 CP7352 CP7350 CP7353 CP7354 CP7355 CP7355 CP7348 CP7353I5 CP7348
AGCP7348EA 714 USD InStock
CAAGCP7353I5 CAAGCP7348 AGCP7348 AGCP7352 AGCP7353 AGCP7354 AGCP7355 AGCP7350 AGCP7351 CAAGCP7355 CAAGCP7354 AGCP7353I5 CAAGCP7351 CAAGCP7350 CAAGCP7353 CAAGCP7352
J&W Porabond PoraBOND Q Columns, Agilent Technologies
Chromatography Columns
An extremely stable bonded porous polymer PLOT stationary phase for analyzing volatile solvents and hydrocarbons.
  • Bonded PLOT column for more reliable results for analysis of volatile solvents and hydrocarbons
  • Extended analysis offers broad application range
  • Temprerature limits of 300 isothermal and 320 °C programmed
  • Engineered for high stability, withstands repeated water injections
  • Proprietary manufacturing technique results in very pure porous polymer with virtually no catalytic activity, allowing operation to 320 °C without decomposition
  • Bonding technology results in greatly reduced particle shedding, reduces the needs for particle traps
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W PoraBOND Q is the long-term solution for analyzing volatile solvents and hydrocarbons. It is the most stable column of its kind and withstands repeated water injections. The porous polymer is very pure and has virtually no catalytic activity. This means that PoraBOND Q can be used up to 320 °C without decomposition.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty