Would you like to log in again?
J&W PLOT PT Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CP7351PT 19091P-K15PT CP7534PT CP7353PT CP7353PT CP7348PT CP7354PT 19091P-K15PT CP7348PT CP7351PT CP7534PT CP7354PT
AGCP7354PTEA 949 USD InStock
AGCP7354PT AGCP7353PT CAAG19091PK15P CAAGCP7354PT AGCP7348PT CAAGCP7348PT AGCP7534PT CAAGCP7534PT AGCP7351PT CAAGCP7353PT CAAGCP7351PT AG19091P-K15PT
J&W PLOT PT Columns, Agilent Technologies
Chromatography Columns
PLOT column with integral particle trap at each end to eliminate the need to connect separate traps, for leak-free GC/MS operation.
  • Enhance analytical capability in the lab using PLOT analyses with MS detection
  • No more hassles with unions or downstream filters, due to integrated particle trapping technology incorporated into the column as one continuous length of fused silica tubing
  • Reproducible flow restriction and increased stability reduce the need for method/system adjustments, and promote a steadier analytical output for higher throughput
  • Lower operating costs and less downtime associated with replacing filters and column switching valves
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W PLOT PT columns have an integral particle trap at each end, eliminating the need to connect separate traps. Operation is more convenient, with no risk of leaks. Integrated particle-trapping technology on both column ends reduces downtime and allows the use of GC/MS for detailed qualitative and quantitative analysis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty