Would you like to log in again?
J&W CP-Wax 51 for Amines Columns, Agilent Technologies
Supplier: Agilent
J&W
CP7405 CP7405
AGCP7405EA 727.8 USD InStock
CAAGCP7405 AGCP7405
J&W CP-Wax 51 for Amines Columns, Agilent Technologies
Chromatography Columns
Agilent J&W CP-Wax 51 for Amines is designed for the analysis of C4 to C10 amines, diamines, and aromatic amines. The column conforms to EPA method 607.
  • Symmetrical peaks due to basic reagent deactivation
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty