Would you like to log in again?
J&W CP-Sil 88 for FAME Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7488 CP7487 CP7489 CP7488 CP7487 CP7489 CP7489I5 CP7489I5
AGCP7487EA 1302 USD InStock
CACP7489 AGCP7487 CAAGCP7487 CAAGCP7489I5 CAAGCP7488 AGCP7488 AGCP7489 AGCP7489I5
J&W CP-Sil 88 for FAME Columns, Agilent Technologies
Chromatography Columns
Guarantees analysis of FAME cis/trans isomers in the C6 to C26 range.
  • Optimized for analysis of FAME cis/trans isomers
  • High-polarity stationary phase increases efficiency and productivity
  • Use for FAME separations in the C6 to C26 range
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Sil 88 for FAME analysis delivers resolution of cis/trans FAME isomers for complete confidence.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty