Would you like to log in again?
J&W CP-Cyclodextrin-β-2,3,6-M-19 Columns, Agilent Technologies
Supplier: Agilent Technologies
CP7501 CP7501
AGCP7501EA 1827 USD InStock
CAAGCP7501 AGCP7501
J&W CP-Cyclodextrin-β-2,3,6-M-19 Columns, Agilent Technologies
Chromatography Columns
Separates many optical isomers that could not be analyzed with previously available methods.
  • Unique selectivity for optical and positional isomer separations
  • High efficiency enables wide range of applications
  • Excellent peak shape for underivatized polar compounds
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Cyclodextrin-β-2,3,6-M-19 separates many optical isomers that were once immune to analysis. Due to its selectivity, o-, m- and p-xylenes can now be separated. The column is also useful for nonchiral compounds. CP-Cyclodextrin-β-2,3,6-M-19 has a very high inertness, enabling separation of underivatized polar compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty