Would you like to log in again?
J&W CP-Chirasil-Dex CB Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7503 CP7502I5 CP7502I5 CP7502 CP7503 CP7502
AGCP7502I5EA 1239 USD InStock
CAAGCP7502I5 AGCP7502I5 CAAGCP7502 CAAGCP7503 AGCP7503 AGCP7502
J&W CP-Chirasil-Dex CB Columns, Agilent Technologies
Chromatography Columns
Delivers the highest resolution between isomers with guaranteed enantioselective stability for improved lifetime.
  • Cyclodextrin bonded to dimethylpolysiloxane for homogeneous enantioselectivity throughout the column
  • High resolution of isomers across a broad application range
  • Chemically bonded phase for excellent longevity
  • No need for derivatization, which improves productivity
  • Low elution temperature of polar compounds
  • Suitable for all injection techniques
  • Similar to LIPODEX C, Rt-β DEXm, β-DEX 110, β-DEX 120
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Chirasil-Dex CB consists of cyclodextrin directly bonded to dimethylpolysiloxane. This prevents the cyclodextrin from migrating to different locations in the surface film, delivering homogeneous enantioselectivity throughout the phase, which provides the highest resolution of isomers. It also guarantees stability of enantioselectivity; as a result, the lifetime of β-cyclodextrin capillary columns is significantly improved. CP-Chirasil-Dex CB permits low elution temperatures of polar compounds and is suitable for all injection techniques.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty