Would you like to log in again?
J&W CP-Molsieve 5Å Columns, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J&W
CP7536 CP7539 CP6938 CP7533 CP7539 CP7538I5 CP7535 CP7544 CP7544 CP7535 CP7543 CP7538 CP7536 CP7538 CP7533 CP6938 CP7538I5 CP7543
AGCP6938EA 1426 USD InStock
CAAGCP7539 CAAGCP7538 CAAGCP7535 CAAGCP7536 AGCP6938 AGCP7536 AGCP7538 AGCP7539 CAAGCP6938 AGCP7543 AGCP7544 AGCP7533 CAAGCP7538I5 AGCP7535 CAAGCP7533 CAAGCP7544 CAAGCP7543 AGCP7538I5
J&W CP-Molsieve 5Å Columns, Agilent Technologies
Chromatography Columns
Delivers highly efficient and accurate separation of argon and oxygen at ambient temperature.
  • Separates argon and oxygen at ambient temperature to reduce costs
  • High efficiency for increased productivity
  • Symmetrical peaks for accurate results
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-Molsieve 5 Å is a molecular-sieve-coated PLOT capillary column specially suited for high-resolution separation of permanent gases. Analysis times are reduced by up to 75% compared to packed columns. Baseline separation of Ar/O₂ is achieved at ambient temperature. The column's thin-layer dimensions produce fast elution of CO with symmetrical peaks.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty