Would you like to log in again?
J&W CP-SimDist Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7571 CP7592 CP7512 CP7532 CP7570 CP7592 CP7582 CP7562 CP7570 CP7569 CP7542 CP7532 CP7582 CP7569 CP7542 CP7512 CP7571 CP7562
AGCP7569EA 836 USD InStock
AGCP7569 AGCP7532 AGCP7512 AGCP7562 AGCP7542 CACP7512 CACP7569 AGCP7592 AGCP7570 AGCP7571 AGCP7582 CAAGCP7542 CAAGCP7532 CAAGCP7582 CAAGCP7571 CAAGCP7592 CAAGCP7570 CAAGCP7562
J&W CP-SimDist Columns, Agilent Technologies
Chromatography Columns
CP-SimDist UltiMetal performs extended analysis to C120, and is guaranteed to a Tmax of 450 °C, due to the virtually unbreakable UltiMetal deactivated stainless steel tubing.
  • Designed for ASTM D2887 and the extended D2887 method compliance
  • Low bleed
  • Extended analysis to C120 with maximum temperature of 450 °C
  • UltiMetal deactivated stainless steel tubing for excellent durability (same id as 0.53 mm id fused silica)
  • Excellent retention time repeatability and column lifetime due to special deactivation of UltiMetal surface
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-SimDist UltiMetal performs extended analysis to C120, and is guaranteed to a maximum temperature of 450 °C, due to the virtually unbreakable inert UltiMetal deactivated stainless steel tubing. The internal diameter is the same as for fused silica 0.53 mm id (Megabore) columns, providing trouble-free automation of on-column injection. The special deactivation applied to the UltiMetal surface gives better retention time repeatability than any other high-temperature column. This column is designed for ASTM D2887 and the extended D2887 method compliance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty