Would you like to log in again?
J&W CP-Sil 8 CB Columns, Agilent Technologies
Supplier: Agilent
CP7771 CP7595 CP7741 CP8511 CP7598 CP7598 CP7696 CP7666 CP7596 CP7696 CP7771 CP7691I5 CP8014 CP6681 CP7595 CP7741 CP7681 CP8511 CP7666 CP8753 CP7691I5 CP7681 CP8751 CP8751 CP7596 CP6681 CP7691 CP8761 CP8761 CP8014 CP8753 CP7691
AGCP7598EA 825.58 USD InStock
CAAGCP7691I5 AGCP7598 AGCP6681 AGCP8761 AGCP7771 AGCP7595 AGCP7596 AGCP7696 AGCP7691 CAAGCP7696 CAAGCP7596 CAAGCP7598 CAAGCP7691 CAAGCP7771 CAAGCP7595 CAAGCP8761 CAAGCP6681 CAAGCP7666 AGCP7741 AGCP8511 AGCP8753 AGCP7666 AGCP8014 AGCP8751 AGCP7681 AGCP7691I5 CAAGCP8751 CAAGCP8014 CAAGCP7741 CAAGCP8511 CAAGCP8753 CAAGCP7681
J&W CP-Sil 8 CB Columns, Agilent Technologies
Chromatography Columns
A general-purpose (5% phenyl) methlypolysiloxane phase that is excellent for method development.
  • (5% phenyl) methylpolysiloxane
  • Nonpolar, general-purpose phase
  • Bonded, crosslinked, solvent rinsable, and low bleed
  • High column-to-column reproducibility
  • Wide range of available dimensions
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SPB-5, Rtx-5, BP-5, OV-5, 007-2(MPS-5), SE-52, SE-54, XTI-5, PTE-5, ZB-5, AT-5, MDN-5, ZB-5Plus
Agilent J&W CP-Sil 8 CB is a general-purpose phase that incorporates 5% phenyl groups in the dimethylpolysiloxane polymer, giving it slightly higher polarity than CP-Sil 5 CB. This delivers better selectivity for aromatic compounds and makes it the best choice for method development. Equivalent to USP G27 and G36, it shows excellent column-to-column reproducibility and very high column efficiencies. UltiMetal columns are recommended for the highest operating temperatures, and when working in rugged environments with process or portable instruments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty