Would you like to log in again?
J&W CP-Wax 57 CB for Glycols and Alcohols Columns, Agilent Technologies
Supplier: Agilent Technologies
CP7615 CP7615 CP7617 CP7617
AGCP7617EA 738 USD InStock
CAAGCP7615 CAAGCP7617 AGCP7617 AGCP7615
J&W CP-Wax 57 CB for Glycols and Alcohols Columns, Agilent Technologies
Chromatography Columns
Optimized for the analysis of glycols, diols, and alcohols.
  • Unique, high polarity wax phase
  • Symmetrical peaks provide the most accurate results
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
The crosslinked and bonded phase delivers robustness and enhanced column lifetime.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty