Would you like to log in again?
J&W CP-Wax 52 CB Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7713II5 CP8763I CP8723I CP8746I CP7658I CP8763I CP7668I CP8553I CP7773I CP7743I CP7791I CP7763I CP8074I CP7658I CP7792I CP8773I CP7773II5 CP8738I CP8746I CP8843I CP7713II5 CP7743I CP7668I CP7773I CP7638I CP8553I CP8773I CP7713I CP7773II5 CP7792I CP7638I CP7791I CP8738I CP8843I CP8723I CP7763I CP8074I CP8713I CP8713I
AGCP7773II5EA 1314 USD InStock
CAAGCP7763I AGCP7773II5 CAAGCP7743I AGCP7792I CAAGCP7773II5 CAAGCP7773I CAAGCP8074I CAAGCP7638I CAAGCP7668I CAAGCP7658I CAAGCP8843I AGCP8723I AGCP8713I AGCP8773I AGCP8763I AGCP8553I CAAGCP8746I AGCP7763I AGCP7773I CAAGCP7792I AGCP8074I AGCP7791I AGCP7638I AGCP7668I AGCP7658I AGCP7713I AGCP7743I CAAGCP8723I AGCP8746I AGCP7713II5 CACP8738I CAAGCP8763I AGCP8738I CAAGCP8773I CAAGCP7713II5 CAAGCP8553I CAAGCP7791I AGCP8843I CAAGCP8713I
J&W CP-Wax 52 CB Columns, Agilent Technologies
Chromatography Columns
Highly polar polyethylene glycol phase that provides separations for a broad range of applications.
  • Polyethylene glycol (PEG) phase
  • High polarity, suitable for a broad range of separations
  • Excellent column-to-column inertness reproducibility
  • Sharp and symmetrical peak shape performance for active polar compounds
  • Bonded, crosslinked, and solvent rinsable
  • Wider temperature range than non-bonded polyethylene glycols
  • High resolution of low boiling point analytes
  • Excellent temperature stability for a variety of EPA and ASTM methods
  • QC tested with highly demanding test probes
  • Performance Summary shipped with each column
  • UltiMetal column recommended for working in rugged environments with process or portable instruments
  • Similar phases: Supelcowax 10, Stabilwax, BP-20, 007-CW, Rtx-WAX, ZB-WAX, ZB-WAXPlus
Agilent J&W CP-Wax 52 CB is a high-polarity polyethylene glycol (PEG) phase with a wider temperature range than nonbonded PEG columns, due to extensive crosslinking. This column delivers higher resolution of low boiling point analytes, with guaranteed reproducibility and excellent temperature stability, and is ideal for EPA and ASTM methods that require PEG columns. The improved line of Agilent J&W CP-Wax 52 CB GC columns utilizes the same innovative inert processes employed in the manufacture of Agilent J&W DB-WAX Ultra Inert columns. These columns have an extended inertness lifetime that withstands repeated temperature cycling to the upper temperature limits of the column, while critical performance parameters such as selectivity, theoretical plates, and retention indices remain unchanged.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty