Would you like to log in again?
J&W Ultimate Plus Deactivated Fused Silica, Agilent Technologies
Supplier: Agilent
J&W
CP802505 CP802530 CP802530 CP803205 CP801510 CP801206 CP805310 CP801805 CP803205 CP801206 CP805305 CP801810 CP803210 CP805330 CP801505 CP803230 CP803210 CP805330 160-7625-5 CP802505 CP801510 CP801805 CP805310 CP805305 CP802510 160-7625-5 CP803230 CP801505 CP802510 CP801810
AGCP801510EA 225.45 USD InStock
AGCP801510 AGCP803210 AGCP805310 AGCP805330 CAAGCP801805 AGCP803230 CAAGCP801206 CACP802505 CAAGCP801505 CAAGCP803205 CAAGCP805305 CAAGCP802530 CAAGCP802510 AGCP801810 AGCP802505 AGCP805305 AGCP802510 AGCP803205 CA160-7625-5 AGCP802530 CAAGCP801510 AG160-7625-5 CAAGCP801810 AGCP801505 CAAGCP803230 CAAGCP805310 CAAGCP803210 CAAGCP805330 AGCP801805 AGCP801206
J&W Ultimate Plus Deactivated Fused Silica, Agilent Technologies
Chromatography Columns
Inert guard column/retention gap used to extend the life of analytical columns when working with heavy matrix samples.
  • Engineered for improved inertness for trace level active compounds
  • For guard columns, retention gaps, restrictors, and transfer lines
  • Particularly useful for for semivolatiles, pesticides, and drugs of abuse applications in heavy sample matrices such as soil, sludge, food, and body fluids
  • Use as guard column to extend life of analytical column
  • Available in a wide variety of inner diameters in lengths up to 30 meters
  • Batch tested
  • Superior replacement for: Siltek, RXI, and Z-guard deactivated tubing
Ultimate Plus Deactivated Fused Silica tubing can be used as guard columns, retention gaps, transfer lines, or restrictors to improve GC flow path performance. It is engineered for optimum inertness and peak shape performance with active trace level compounds, for analysts in environmental, food safety, and forensic labs running heavy matrix samples. For optimum system performance, use Ultimate Plus Deactivated Fused Silica with Ultra Inert GC columns, Ultra Inert liners, and Ultra Inert gold seals. Ensure leak-free GC connections with Ultra Inert treated press fit unions or Ultimate Unions and flexible metal ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty