Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

J&W VF-35ms Columns, Agilent Technologies

Supplier: Agilent Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

J&W
CP8884 CP8877 CP8874 CP8877 CP8884 CP8874
AGCP8884EA 857 USD InStock
AGCP8884 CAAGCP8877 CAAGCP8884 CAAGCP8874 AGCP8874 AGCP8877
J&W VF-35ms Columns, Agilent Technologies
Chromatography Columns
Has a medium polarity stationary phase and is ideal for trace environmental and chemical analyses, and as a confirmation column.

  • Stabilized arylene-modified equivalent of a 35% phenylmethyl phase
  • Ideal for dual-column confirmational analyses
  • Medium polarity ideal for trace environmental and chemical analyses
  • 0.15 mm Int. Ø columns available for high efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column

The VF-35ms is a low-bleed, highly stable column with a programmable maximum temperature of 360 °C.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty