Would you like to log in again?
VF-1ms Columns, Agilent Technologies
Supplier: Agilent
J&W
CP8904 CP8930 CP8907 CP8916 CP8912 CP8930 CP8913 CP8916 CP8913 CP8907 CP8904 CP8912
AGCP8916EA 1379.42 USD InStock
CAAGCP8913 CAAGCP8916 CAAGCP8904 AGCP8916 AGCP8904 AGCP8907 CAAGCP8912 AGCP8912 CAAGCP8930 AGCP8913 AGCP8930 CAAGCP8907
VF-1ms Columns, Agilent Technologies
Chromatography Columns
A highly inert, nonpolar, low-bleed, PDMS phase, ideal for GC/MS and for trace analysis in a wide range of applications.
  • Highly inert, nonpolar, 100% dimethylpolysiloxane phase
  • Low-bleed GC column providing increased sensitivity over a broad range of applications
  • QC test results for retention index, efficiency, selectivity, and bleed reported with every column
  • 0.15 mm Int.Ø columns available for high efficiency GC and GC/MS analysis
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-1ms is a highly inert, nonpolar, 100% dimethylpolysiloxane phase column that provides increased sensitivity for a broad array of applications, particularly for energy, pharmaceutical, and environmental analysis. The column has an ultra-low-bleed specification of 1 pA at 325 °C (30 m, 0.25 mm, 0.25 μm) for trace analysis with GC/MS.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty