Would you like to log in again?
J&W 8890 GC Columns with Smart Key, Agilent Technologies
Supplier: Agilent
J&W
122-5532UI-KEY 122-7032-KEY 122-5532UI-KEY 125-7032-KEY 122-5032-KEY 19091S-431UI-KEY 19091S-431UI-KEY 122-5532-KEY CP8944-KEY CP8944-KEY 122-7032-KEY 122-5032-KEY 19091J-413-KEY 19091S-433-KEY 19091J-413-KEY 125-7032-KEY 122-5532-KEY 19091S-433-KEY
AG122-5532-KEYEA 850.29 USD InStock
CAAG1225032KEY CAAGCP8944-KEY CAAG19091S433K CAAG1225532UIK CAAG19091S43UK AG122-5532-KEY CAAG19091J413K AGCP8944-KEY CAAG1225532KEY AG19091S433KEY AG1225532UIKEY AG122-5032-KEY AG122-7032-KEY AG19091J413KEY AG1909S431UKEY CAAG1257032KEY AG125-7032-KEY CAAG1227032KEY
J&W 8890 GC Columns with Smart Key, Agilent Technologies
Chromatography Columns
Various GC columns with a Smart Key included, made specifically to fit the 8890 GC.
  • Smart Key allows self-configuration, usage monitoring, and help automating method development when using 8890 GC
  • Wide selection of available stationary phases (low, mid, and high polarity, application-specific stationary phases)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent 8890 GC system
Agilent J&W offers a selection of GC columns with Smart Key (compatible only with the 8890 GC). Smart Keys allow for immediate identification and usage monitoring of your GC column. A variety of stationary phases are available, ranging from Ultra Inert and Poly Ethylene Glycol (PEG) to unique, application-specific stationary phases. If your configuration is not available, please request a custom column with Smart Key through the Custom Column Shop.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty