Would you like to log in again?
J&W EZ-Guard Columns, Agilent Technologies
Supplier: Agilent Technologies
CP9014 CP9011 CP9013 CP9010 CP9013 CP9021 CP9023 CP9011 CP9023 CP9014 CP9021 CP9010 CP9012 CP9012
AGCP9014EA 890 USD InStock
AGCP9014 CAAGCP9010 AGCP9013 CAAGCP9021 AGCP9021 AGCP9010 AGCP9023 AGCP9012 AGCP9011 CAAGCP9013 CAAGCP9014 CAAGCP9011 CAAGCP9023 CAAGCP9012
J&W EZ-Guard Columns, Agilent Technologies
Chromatography Columns
Combines a VF column with a built-in guard column to avoid problems associated with unions.
  • Built-in guard columns eliminate the need for press-fit connectors
  • Minimizes front-end contamination and increases column lifetime
  • Improves focusing sample onto the front of the column for better peak shape
  • Minimizes MSD contamination originating from the column (when used as transfer line to the MS detector)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
EZ-Guard columns combine an Agilent J&W VF column with a built-in guard column. The first 5 or 10 m section of the column (depending on the configuration) is not coated with stationary phase, but has been deactivated. Eliminating the column connection between the guard and analytical sections ensures 100% leak-free performance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty