Would you like to log in again?
J&W VF-5ms Columns, Agilent Technologies
Supplier: Agilent
CP8962 CP8945 CP9043 CP8944 CP8962 CP9034 CP8944 CP8939 CP8946 CP9043 CP9034 CP8957 CP8960 CP8960 CP8946 CP8939 CP8957 CP8945
AGCP8939EA 495.14 USD InStock
CAAGCP8946 CAAGCP8957 CAAGCP8945 AGCP8939 AGCP8945 AGCP8944 AGCP8946 AGCP8957 AGCP8962 CAAGCP8939 CAAGCP9043 AGCP8960 AGCP9043 CACP8944 AGCP9034 CAAGCP8960 CAAGCP8962 CAAGCP9034
J&W VF-5ms Columns, Agilent Technologies
Chromatography Columns
An inert, low-bleed 5% phenylmethyl polysiloxane phase for increased sensitivity, accuracy, and instrument uptime for semipolar and polar compounds.
  • Highly inert 5% phenylmethyl polysiloxane column for increased sensitivity, accuracy, and instrument uptime
  • Minimal column bleed improves sensitivity (ultra-low bleed specification of 1 pA @ 325 °C (for 30000×0.25 mm, 0.25 μm))
  • Slightly higher polarity than VF-1ms for improved selectivity for aromatic compounds
  • Excellent selectivity and inertness, applicable to a wide range of semipolar and even polar compounds
  • 0.15 mm id columns available for high efficiency GC and GC/MS analyses
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-5ms is an inert 5% phenylmethyl polysiloxane column that delivers increased sensitivity, accuracy, and instrument uptime. The column has the lowest guaranteed bleed specification of 1 pA at 325 °C (for 30 m, 0.25 mm, 0.25 µm). This column has slightly higher polarity than VF-1ms, resulting in better selectivity for aromatic compounds. This selectivity, combined with superior inertness, makes these columns applicable to a wide range of semipolar and even polar compounds, such as phenols.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty