Would you like to log in again?
J&W VF-5 Pesticide Columns, Agilent Technologies
Supplier: Agilent Technologies
CP9074 CP9074
AGCP9074EA 843 USD InStock
CAAGCP9074 AGCP9074
J&W VF-5 Pesticide Columns, Agilent Technologies
Chromatography Columns
Specially designed for the determination of trace levels of pesticide residues with ECD or MS detection.
  • Specially designed for the determination of trace levels of pesticide residue
  • Highly inert for enhanced ECD and MS detection
  • Tested with key pesticides, including endrin and aldrin, for optimal performance and consistency of results
  • Low bleed
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-5 Pesticide column is specially designed for the determination of trace levels of pesticide residue. It is highly inert for enhanced ECD and MS detection. Every column is individually tested before shipment with key pesticides, including endrin and aldrin, ensuring optimal performance and consistency of results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty