Would you like to log in again?
J&W Select Biodiesel Columns, Agilent Technologies
Supplier: Agilent
J&W
CP9076 CP9076 CP9078 CP9078
AGCP9076EA 886.32 USD InStock
CAAGCP9076 AGCP9076 AGCP9078 CAAGCP9078
J&W Select Biodiesel Columns, Agilent Technologies
Chromatography Columns
Good column lifetime when operating at very high temperature up to 400 °C with ASTM and EN biodiesel methods.
  • Complete set of biodiesel columns for full compliance and ease-of-use
  • UltiMetal deactivation technology for stainless steel tubing provides high accuracy and longevity
  • Pre-tested for complete confidence in results
  • Convenient pre-coupled retention gap that is leak tested
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Although traditional fused silica can be used, high temperatures often mean shortened column lifetimes. Using an UltiMetal deactivated stainless steel column with an ultra-stable stationary phase produces more consistent results and virtually eliminates column breakage.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty