Would you like to log in again?
J&W VF-5ht Columns, Agilent Technologies
Supplier: Agilent Technologies
CP9094 CP9047 CP9046 CP9047 CP9094 CP9046
AGCP9047EA 543 USD InStock
AGCP9047 AGCP9046 CAAGCP9094 AGCP9094 CAAGCP9046 CAAGCP9047
J&W VF-5ht Columns, Agilent Technologies
Chromatography Columns
VF-5ht improves the analysis of high boiling compounds by exhibiting low bleed, even at high temperatures.
  • Enhanced stability improves column longevity and reduces downtime
  • Superior detector performance provides improved detection limits
  • Ultra low bleed at high temperatures for analysis of high-boiling compounds
  • Optimized sensitivity and accuracy for analysis of high molecular weight compounds
  • Identical selectivity as VF-5ms (bleed spec of 30 m × 0.25 mm column is < 5 pA at 400 °C)
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W VF-5ht improves the analysis of high boiling compounds with low-bleed performance, even at high temperatures. Based on ultra-low-bleed technology, VF-5ht provides unmatched selectivity, sensitivity, and accuracy for the analysis of high molecular weight compounds. This column has temperature limits of –60 to 400 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty